4-Nitrophenyl hexanoate 95%

Supplier: Ambeed
Warning

Synonyms: Hexanoic Acid 4-Nitrophenyl Ester

A955912-10G A955912-1G A955912-5G
76993-996EA 39 USD
76993-996 76993-998 76994-000
4-Nitrophenyl hexanoate 95%
4-Nitrophenyl hexanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00056111
CAS Number: 956-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR