tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A955783-100MG A955783-10G A955783-1G A955783-250MG A955783-25G A955783-5G
77037-008EA 19.1 USD
77037-008 77037-010 77037-012 77037-014 77037-016 77037-018
tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylate 95%
tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₃
MW: 202.25 g/mol
MDL Number: MFCD14705159
CAS Number: 190141-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR