4-Bromo-2-tert-butylphenol 95%

Supplier: Ambeed
Warning

A953117-100G A953117-10G A953117-1G A953117-5G
76993-980EA 360 USD
76993-980 76993-982 76993-984 76993-986
4-Bromo-2-tert-butylphenol 95%
4-Bromo-2-tert-butylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
MDL Number: MFCD00192645
CAS Number: 10323-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR