2-Hydroxy-3-iodobenzaldehyde 98%

Supplier: Ambeed
Warning

A952513-250MG A952513-1G A952513-100MG
77037-006EA 38 USD
77037-006 77670-390 77873-386
2-Hydroxy-3-iodobenzaldehyde 98%
2-Hydroxy-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅IO₂
MDL Number: MFCD13180433
CAS Number: 23602-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR