4-Fluoro-3-pyrrolidinobenzaldehyde 98%

Supplier: Ambeed
Warning

A951669-100MG A951669-1G A951669-250MG
77037-000EA 31 USD
77037-000 77037-002 77037-004
4-Fluoro-3-pyrrolidinobenzaldehyde 98%
4-Fluoro-3-pyrrolidinobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO
MDL Number: MFCD13191603
CAS Number: 1197193-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR