[3-(Aminomethyl)phenyl]methanol 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminomethyl)benzyl Alcohol

A951663-10G A951663-250MG A951663-1G A951663-5G
77036-996EA 264 USD
77036-996 77036-998 77603-344 77670-384
[3-(Aminomethyl)phenyl]methanol 97%
[3-(Aminomethyl)phenyl]methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD06212981
CAS Number: 34231-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR