2,5-Dimethylthiophene-3-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2,5-Dimethylthiophen-3-yl)boronic acid

A951039-100MG A951039-1G A951039-250MG A951039-5G
77020-070EA 33.95 USD
77020-070 77020-072 77020-074 77020-076
2,5-Dimethylthiophene-3-boronic acid 95%
2,5-Dimethylthiophene-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₉BO₂S
MW: 156.01 g/mol
MDL Number: MFCD09837620
CAS Number: 162607-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR