2-(Cyclohexylamino)ethanesulphonic acid (CHES) 98%

Supplier: Ambeed
Warning

Synonyms: N-Cyclohexyltaurine, CHES

A950389-100G A950389-25G A950389-500G A950389-5G A950389-1000G
77036-988EA 26 USD
77036-988 77036-990 77036-992 77036-994 77674-414
2-(Cyclohexylamino)ethanesulphonic acid (CHES) 98%
2-(Cyclohexylamino)ethanesulphonic acid (CHES)

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃S
MDL Number: MFCD00003835
CAS Number: 103-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR