4-((Trifluoromethyl)thio)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
A950109-100G
A950109-1G
A950109-25G
A950109-5G
77007-170EA
113
USD
77007-170
77007-172
77007-174
77007-176
4-((Trifluoromethyl)thio)benzoic acid 98%
4-((Trifluoromethyl)thio)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₅F₃O₂S |
MDL Number:
MFCD00040906 CAS Number: 330-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...