1,3-Thiazole-4-carbohydrazide 98%

Supplier: Ambeed
Warning

Synonyms: Thiazole-4-carbohydrazide

A944889-1G A944889-250MG
77020-060EA 93.36 USD
77020-060 77020-062
1,3-Thiazole-4-carbohydrazide 98%
1,3-Thiazole-4-carbohydrazide

Precaución: for Research Use Only

Formula: C₄H₅N₃OS
MW: 143.17 g/mol
MDL Number: MFCD00614363
CAS Number: 101257-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR