2-Amino-4-[(3,4-diaminophenyl)sulfonyl]phenylamine 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
4,4'-sulphonylbis(benzene-1,2-diamine)
A944432-1G
A944432-5G
77007-154EA
216.43
USD
77007-154
77007-156
2-Amino-4-[(3,4-diaminophenyl)sulfonyl]phenylamine 95%
2-Amino-4-[(3,4-diaminophenyl)sulfonyl]phenylamine
Precaución: for Research Use Only
Formula:
C₁₂H₁₄N₄O₂S MW: 278.33 g/mol |
MDL Number:
MFCD00437381 CAS Number: 13224-79-8 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...