3-Bromo-6-chloroimidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Warning

A943810-1G A943810-250MG A943810-5G
77020-050EA 23 USD
77020-050 77020-052 77891-710
3-Bromo-6-chloroimidazo[1,2-a]pyridine 98%
3-Bromo-6-chloroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MDL Number: MFCD08667858
CAS Number: 886371-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR