Methyl-4-(4-aminophenoxy)benzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-(4-aminophenoxy)benzoate
A943333-1G
A943333-250MG
77007-150EA
356.47
USD
77007-150
77007-152
Methyl-4-(4-aminophenoxy)benzoate 98%
Methyl-4-(4-aminophenoxy)benzoate
Precaución: for Research Use Only
Formula:
C₁₄H₁₃NO₃ MW: 243.26 g/mol |
MDL Number:
MFCD00445273 CAS Number: 24477-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...