3-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

A942108-10G A942108-1G A942108-25G A942108-5G
77020-042EA 118.83 USD
77020-042 77020-044 77020-046 77020-048
3-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester 97%
3-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₄BNO₄S
MW: 373.27 g/mol
MDL Number: MFCD05663863
CAS Number: 796061-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR