Ethyl 3-((tert-butoxycarbonyl)amino)propanoate 95%

Supplier: Ambeed
Warning

A939830-100MG A939830-1G
77020-030EA 12.74 USD
77020-030 77020-032
Ethyl 3-((tert-butoxycarbonyl)amino)propanoate 95%
Ethyl 3-((tert-butoxycarbonyl)amino)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
MDL Number: MFCD10000637
CAS Number: 88574-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR