3-Bromo-5-methylpyrazin-2(1H)-one 95%

Supplier: Ambeed
Warning

A938986-100MG A938986-250MG
76993-962EA 61 USD
76993-962 76993-964
3-Bromo-5-methylpyrazin-2(1H)-one 95%
3-Bromo-5-methylpyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MDL Number: MFCD13193675
CAS Number: 1260810-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR