1-Phenylpyrrolidin-3-amine 95%

Supplier: Ambeed
Warning

A938850-1G A938850-100MG
77020-028EA 308 USD
77020-028 77891-704
1-Phenylpyrrolidin-3-amine 95%
1-Phenylpyrrolidin-3-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂
MDL Number: MFCD11640906
CAS Number: 18471-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR