4-Fluoro-3-morpholinobenzaldehyde 98%

Supplier: Ambeed
Warning

A938038-100MG A938038-1G A938038-250MG
77036-974EA 25.47 USD
77036-974 77036-976 77036-978
4-Fluoro-3-morpholinobenzaldehyde 98%
4-Fluoro-3-morpholinobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₂
MW: 209.2169 g/mol
MDL Number: MFCD13191604
CAS Number: 1197193-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR