(3-(2-Methoxyethoxy)phenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A936787-1G
A936787-250MG
A936787-5G
77020-020EA
114.58
USD
77020-020
77020-022
77020-024
(3-(2-Methoxyethoxy)phenyl)boronic acid 98%
(3-(2-Methoxyethoxy)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₃BO₄ MW: 196.01 g/mol |
MDL Number:
MFCD11183195 CAS Number: 227305-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...