6-Bromo-2,3-difluorophenol 97%

Supplier: Ambeed
Warning

A935644-100MG A935644-1G A935644-250MG
77036-964EA 40.32 USD
77036-964 77036-966 77036-968
6-Bromo-2,3-difluorophenol 97%
6-Bromo-2,3-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MW: 208.988 g/mol
MDL Number: MFCD11656842
CAS Number: 186590-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR