2-(4-Bromophenyl)-6-chloroquinoline-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A935239-1G A935239-250MG A935239-5G
77007-146EA 134 USD
77007-146 77007-148 77882-837
2-(4-Bromophenyl)-6-chloroquinoline-4-carboxylic acid 97%
2-(4-Bromophenyl)-6-chloroquinoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₉BrClNO₂
MDL Number: MFCD02319315
CAS Number: 342017-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR