5-(Methylsulfonyl)pyridin-2-amine 98%

Supplier: Ambeed
Warning

A934526-100MG A934526-1G A934526-250MG
77020-012EA 84.88 USD
77020-012 77020-014 77020-016
5-(Methylsulfonyl)pyridin-2-amine 98%
5-(Methylsulfonyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
MW: 172.2 g/mol
MDL Number: MFCD09946387
CAS Number: 35196-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR