(S)-4-(tert-Butyl)-2-(2-(diphenylphosphino)phenyl)-4,5-dihydrooxazole 98_ee99, Ambeed.Inc

Supplier: Ambeed
Warning

A934451-1G A934451-250MG A934451-100MG A934451-5G
76993-946EA 186 USD
76993-946 77552-736 77569-660 77684-884
(S)-4-(tert-Butyl)-2-(2-(diphenylphosphino)phenyl)-4,5-dihydrooxazole 98_ee99, Ambeed.Inc
(S)-4-(tert-Butyl)-2-(2-(diphenylphosphino)phenyl)-4,5-dihydrooxazole

Precaución: For research use only.

Formula: C₂₅H₂₆NOP
MDL Number: MFCD09998277
CAS Number: 148461-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR