1,3,5-Tri(p-tolyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-Tris(4-Methylphenyl)benzene, 4,4''-Dimethyl-5'-(p-tolyl)-1,1':3',1''-terphenyl

A934075-1G A934075-25G A934075-5G
76993-940EA 9 USD
76993-940 76993-942 76993-944
1,3,5-Tri(p-tolyl)benzene 98%
1,3,5-Tri(p-tolyl)benzene

Precaución: for Research Use Only

Formula: C₂₇H₂₄
MW: 348.49 g/mol
MDL Number: MFCD00192798
CAS Number: 50446-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR