2-(Bromomethyl)-4-fluorophenylboronic acid neopentyl glycol ester 98%

Supplier: Ambeed
Danger

Synonyms: 2-(2-(Bromomethyl)-4-fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane

A932950-100MG A932950-1G A932950-250MG
77020-006EA 36.08 USD
77020-006 77020-008 77020-010
2-(Bromomethyl)-4-fluorophenylboronic acid neopentyl glycol ester 98%
2-(Bromomethyl)-4-fluorophenylboronic acid neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BBrFO₂
MW: 300.96 g/mol
MDL Number: MFCD06659913
CAS Number: 673456-16-1
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR