4-Fluoro-N,N-dimethylbenzamide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N,N-Dimethyl 4-fluorobenzamide
A932453-10G
A932453-5G
A932453-1G
76993-926EA
228.1
USD
76993-926
76993-928
77684-883
4-Fluoro-N,N-dimethylbenzamide 97%
4-Fluoro-N,N-dimethylbenzamide
Precaución: For research use only.
Formula:
C₉H₁₀FNO MW: 167.18 g/mol |
MDL Number:
MFCD01214152 CAS Number: 24167-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...