5-Bromo-N-methylpyridin-3-amine 95%

Supplier: Ambeed
Warning

A931609-100MG A931609-250MG
76993-924EA 19 USD
76993-924 77891-698
5-Bromo-N-methylpyridin-3-amine 95%
5-Bromo-N-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MDL Number: MFCD13190575
CAS Number: 873383-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR