3,3'-Dibromo-5,5'-bis(trimethylsilyl)-2,2'-bithiophene 95%

Supplier: Ambeed
Warning

A930136-100MG A930136-1G A930136-250MG A930136-5G
77036-958EA 17 USD
77036-958 77036-960 77873-377 77873-378
3,3'-Dibromo-5,5'-bis(trimethylsilyl)-2,2'-bithiophene 95%
3,3'-Dibromo-5,5'-bis(trimethylsilyl)-2,2'-bithiophene

Precaución: for Research Use Only

Formula: C₁₄H₂₀Br₂S₂Si₂
MDL Number: MFCD16621088
CAS Number: 207742-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR