5-Bromo-1-methyl-1H-indole-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A929545-100MG A929545-250MG A929545-5G A929545-1G
77036-952EA 15.92 USD
77036-952 77036-954 77036-956 77603-312
5-Bromo-1-methyl-1H-indole-3-carbaldehyde 95%
5-Bromo-1-methyl-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MDL Number: MFCD03906373
CAS Number: 10102-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR