6-Bromo-3-methoxy-1-phenyl-1H-indazole 95%

Supplier: Ambeed
Warning

A928209-100MG A928209-250MG A928209-1G
77019-996EA 82.76 USD
77019-996 77019-998 77595-500
6-Bromo-3-methoxy-1-phenyl-1H-indazole 95%
6-Bromo-3-methoxy-1-phenyl-1H-indazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂O
MW: 303.15 g/mol
MDL Number: MFCD27939304
CAS Number: 1332527-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR