4-Bromo-3-hydroxybenzamide 95%

Supplier: Ambeed
Warning

A928202-1G A928202-250MG
77019-992EA 37 USD
77019-992 77019-994
4-Bromo-3-hydroxybenzamide 95%
4-Bromo-3-hydroxybenzamide

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD16999309
CAS Number: 916213-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR