(3R)-Morpholine-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A926667-100MG
A926667-1G
A926667-5G
77019-988EA
13
USD
77019-988
77019-990
77891-695
(3R)-Morpholine-3-carboxylic acid 95%
(3R)-Morpholine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₉NO₃ |
MDL Number:
MFCD03426306 CAS Number: 106825-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...