Methyl 3-bromo-4-(methylsulfonyl)benzoate 98%

Supplier: Ambeed
Warning

A926637-1G A926637-250MG A926637-5G
77019-982EA 125.19 USD
77019-982 77019-984 77019-986
Methyl 3-bromo-4-(methylsulfonyl)benzoate 98%
Methyl 3-bromo-4-(methylsulfonyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₄S
MW: 293.13 g/mol
MDL Number: MFCD18651679
CAS Number: 182003-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR