(S)-Methyl-2-amino-3-(1-trityl-1H-imidazol-4-yl)propanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-His(Trt)-OMe.HCl, H-His(Trt)-OMe·HCl, H-His(1-Trt)-OMe·HCl
A926345-100G
A926345-10G
A926345-1G
A926345-250MG
A926345-25G
A926345-5G
77007-128EA
587.74
USD
77007-128
77007-130
77007-132
77007-134
77007-136
77007-138
(S)-Methyl-2-amino-3-(1-trityl-1H-imidazol-4-yl)propanoate hydrochloride 97%
(S)-Methyl-2-amino-3-(1-trityl-1H-imidazol-4-yl)propanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₂₆H₂₅N₃O₂·HCl MW: 447.963 g/mol |
MDL Number:
MFCD00237100 CAS Number: 32946-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...