(R)-7,8-Difluorochroman-4-amine 95%

Supplier: Ambeed
Warning

A926145-100MG A926145-250MG
77036-948EA 141.1 USD
77036-948 77036-950
(R)-7,8-Difluorochroman-4-amine 95%
(R)-7,8-Difluorochroman-4-amine

Precaución: for Research Use Only

Formula: C₉H₉F₂NO
MW: 185.17 g/mol
MDL Number: MFCD07374043
CAS Number: 1213550-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR