2-Methyl-2H-indazole-4-carboxylic acid methyl ester 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methyl-2H-indazole-4-carboxylate

A925210-100MG A925210-1G A925210-250MG
77036-940EA 22.28 USD
77036-940 77036-942 77036-944
2-Methyl-2H-indazole-4-carboxylic acid methyl ester 96%
2-Methyl-2H-indazole-4-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
MDL Number: MFCD11109405
CAS Number: 1071428-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR