(1-Methylpiperidin-4-yl)acetic acid 98%

Supplier: Ambeed
Warning

A925186-100MG A925186-1G A925186-250MG A925186-5G
77036-932EA 14 USD
77036-932 77036-934 77036-936 77036-938
(1-Methylpiperidin-4-yl)acetic acid 98%
(1-Methylpiperidin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MDL Number: MFCD09055305
CAS Number: 87647-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR