2-Bromo-6-chlorophenylboronic acid 98%

Supplier: Ambeed
Warning

A924637-1G A924637-250MG
77007-122EA 45 USD
77007-122 77007-124
2-Bromo-6-chlorophenylboronic acid 98%
2-Bromo-6-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrClO₂
MDL Number: MFCD08701777
CAS Number: 1107580-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR