2-Bromo-4-fluoro-5-methoxyaniline 95%

Supplier: Ambeed
Warning

A924416-100MG A924416-1G A924416-250MG A924416-5G
76993-904EA 29.71 USD
76993-904 76993-906 76993-908 76993-910
2-Bromo-4-fluoro-5-methoxyaniline 95%
2-Bromo-4-fluoro-5-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFNO
MW: 220.03 g/mol
MDL Number: MFCD09800756
CAS Number: 420786-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR