Methyl isoindoline-5-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A924205-100MG A924205-1G A924205-250MG A924205-5G
77036-924EA 29 USD
77036-924 77036-926 77036-928 77036-930
Methyl isoindoline-5-carboxylate hydrochloride 98%
Methyl isoindoline-5-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂
MW: 213.66 g/mol
MDL Number: MFCD10699381
CAS Number: 127168-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR