Methyl-6-chloro-5-iodonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-5-iodonicotinate

A923878-100MG A923878-1G A923878-250MG A923878-5G
77007-112EA 45.62 USD
77007-112 77007-114 77007-116 77007-118
Methyl-6-chloro-5-iodonicotinate 98%
Methyl-6-chloro-5-iodonicotinate

Precaución: for Research Use Only

Formula: C₇H₅ClINO₂
MW: 297.48 g/mol
MDL Number: MFCD08276963
CAS Number: 365413-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR