2-(4-Bromo-3-chlorophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A923721-1G A923721-25G
76993-902EA 101.85 USD
76993-902 77684-878
2-(4-Bromo-3-chlorophenyl)acetic acid 95%
2-(4-Bromo-3-chlorophenyl)acetic acid

Precaución: For research use only.

Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
MDL Number: MFCD18390583
CAS Number: 1261643-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR