tert-Butyl ((1S,3S)-3-hydroxycyclopentyl)carbamate 97%

Supplier: Ambeed
Warning

A922055-250MG A922055-1G A922055-100MG A922055-5G
77036-916EA 42 USD
77036-916 77595-490 77873-370 77873-371
tert-Butyl ((1S,3S)-3-hydroxycyclopentyl)carbamate 97%
tert-Butyl ((1S,3S)-3-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD11226224
CAS Number: 154737-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR