4-Phenoxyaniline 98%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-4-phenoxybenzol

A921564-25G A921564-500G A921564-5G
77007-106EA 10.61 USD
77007-106 77007-108 77007-110
4-Phenoxyaniline 98%
4-Phenoxyaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
MDL Number: MFCD00007862
CAS Number: 139-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR