5-Bromo-2-isopropoxypyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-isoproxypyrimidine

A920473-1G A920473-250MG A920473-25G A920473-5G
76993-882EA 76.39 USD
76993-882 76993-884 76993-886 76993-888
5-Bromo-2-isopropoxypyrimidine 98%
5-Bromo-2-isopropoxypyrimidine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O
MW: 217.06 g/mol
MDL Number: MFCD12024885
CAS Number: 121487-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR