2-(Boc-amino)-2-(2-naphthyl)acetic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((tert-Butoxycarbonyl)amino)-2-(naphthalen-2-yl)acetic acid
A919505-100MG
A919505-1G
A919505-250MG
77019-960EA
21
USD
77019-960
77019-962
77019-964
2-(Boc-amino)-2-(2-naphthyl)acetic acid 95%
2-(Boc-amino)-2-(2-naphthyl)acetic acid
Precaución: for Research Use Only
Formula:
C₁₇H₁₉NO₄ |
MDL Number:
MFCD02683164 CAS Number: 33741-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...