(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, (3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinylphosphonium hexafluorophosphate, ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), PyAOP, (3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinylphosphorous hexafluorophosphate
A919389-10G
A919389-1G
A919389-25G
A919389-100G
A919389-5G
A919389-500G
77007-094EA
22
USD
77007-094
77007-096
77007-098
77569-640
77573-840
77674-364
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Precaución: For research use only.
Formula:
C₁₇H₂₇F₆N₇OP₂ MW: 521.4 g/mol |
MDL Number:
MFCD03703417 CAS Number: 156311-83-0 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...