Benzyl-6-bromohexyl ether 97%

Supplier: Ambeed
Warning

Synonyms: (((6-Bromohexyl)oxy)methyl)benzene, Benzyl 6-Bromohexyl Ether, Benzyloxyhexyl Bromide, (6-Bromohexyloxymethyl)benzene

A917468-1G A917468-25G A917468-5G A917468-100G
77019-954EA 19.1 USD
77019-954 77019-956 77019-958 77684-873
Benzyl-6-bromohexyl ether 97%
Benzyl-6-bromohexyl ether

Precaución: For research use only.

Formula: C₁₃H₁₉BrO
MW: 271.20 g/mol
MDL Number: MFCD02258455
CAS Number: 54247-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR