2-Undecanone 98%

Supplier: Ambeed
Danger

Synonyms: Methyl nonyl ketone, Nonyl methyl ketone

A917403-25G A917403-100G A917403-500G
77007-082EA 8.49 USD
77007-082 77603-302 77674-360
2-Undecanone 98%
2-Undecanone

Precaución: for Research Use Only

Formula: C₁₁H₂₂O
MW: 170.3 g/mol
MDL Number: MFCD00009583
CAS Number: 112-12-9
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR