3,4-Dichlorobenzylamine 95%

Supplier: Ambeed
Danger

A917276-1G A917276-5G A917276-10G
77036-882EA 234.46 USD
77036-882 77036-884 77036-886 77882-821
3,4-Dichlorobenzylamine 95%
3,4-Dichlorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N
MDL Number: MFCD00008114
CAS Number: 102-49-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR